missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Meckelin Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-03802-100UL
This item is not returnable.
View return policy
Description
Meckelin Polyclonal specifically detects Meckelin in Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Meckelin | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 91147 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| meckel syndrome type 3 protein, Meckelin, MGC26979, TNEM67, transmembrane protein 67 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 780-940 of human Meckelin (NP_714915.3). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction