missing translation for 'onlineSavingsMsg'
Learn More

Mebendazole, TRC

Product Code. 30136123
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136123 50 g 50g
30138655 10 g 10g
17962185 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136123 Supplier Toronto Research Chemicals Supplier No. TRCM20050050G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31431-39-7
Molecular Formula C16 H13 N3 O3
Molecular Weight (g/mol) 295.29
Synonym Carbamic acid, N-(6-benzoyl-1H-benzimidazol-2-yl)-, methyl ester, 2-Benzimidazolecarbamic acid, 5-benzoyl-, methyl ester (8CI), Carbamic acid, (5-benzoyl-1H-benzimidazol-2-yl)-, methyl ester (9CI), (5-Benzoyl-1H-benzimidazol-2-yl)carbamic acid methyl ester, 5-Benzoyl-2-benzimidazolecarbamic acid methyl ester, Bantenol, Besantin, Equivurm Plus, Lomper, Mebendazole, Mebenvet, Mebex, Methyl 5-benzoyl-2-benzimidazolecarbamate, Methyl 5-benzoyl-2-benzimidazolylcarbamate, NSC 184849, Noverme, Ovitelmin, Pantelmin, R 17635, Telmin, Vermicidin, Vermirax, Vermox, Verpanyl, Zhihuanqing
IUPAC Name methyl N-(5-benzoyl-1H-benzimidazol-2-yl)carbamate
SMILES COC(=O)Nc1nc2cc(ccc2[nH]1)C(=O)c3ccccc3

Specifications

Quantity 50 g
InChI Formula InChI=1S/C16H13N3O3/c1-22-16(21)19-15-17-12-8-7-11(9-13(12)18-15)14(20)10-5-3-2-4-6-10/h2-9H,1H3,(H2,17,18,19,21)
Formula Weight 295.0957
Percent Purity >95%
Chemical Name or Material Mebendazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.