missing translation for 'onlineSavingsMsg'
Learn More

Mebendazole

Product Code. 15723447 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15723447 5 g 5g
15733447 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15723447 Supplier Thermo Scientific Acros Supplier No. 101720050
  • ONLINE EXCLUSIVE PRICE
    £27.30
    £24.65 / 5g
    Save £2.65
    9% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 31431-39-7 | C16H13N3O3 | 295.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31431-39-7
Molecular Formula C16H13N3O3
Molecular Weight (g/mol) 295.30
MDL Number MFCD00057872
InChI Key OPXLLQIJSORQAM-UHFFFAOYSA-N
Synonym mebendazole, vermox, telmin, mebenvet, pantelmin, vermirax, mebenoazole, ovitelmin, bantenol, mebutar
PubChem CID 4030
ChEBI CHEBI:6704
IUPAC Name methyl N-(6-benzoyl-1H-benzimidazol-2-yl)carbamate
SMILES COC(=O)NC1=NC2=CC=C(C=C2N1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 287.0°C
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in dmso, insoluble in alcohols
Formula Weight 295.29
Chemical Name or Material Mebendazole
compliance-icons
Product Identifier
  • Mebendazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Mebendazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.