missing translation for 'onlineSavingsMsg'
Learn More

Mebendazole, MedChemExpress

Product Code. 16401709
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16401709 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16401709 Supplier Medchem Express Supplier No. HY17595
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Mebendazole is a highly effective, broad-spectrum antihelmintic indicated for the treatment of nematode infestations; has been found as a hedgehog inhibitor.

Mebendazole is a highly effective, broad-spectrum antihelmintic indicated for the treatment of nematode infestations

Specifications

Chemical Name or Material Mebendazole
CAS 31431-39-7
Color White
Purity Grade Notes Research
Molecular Formula C16H13N3O3
Quantity 1 g
Solubility Information DMSO : 4.17 mg/mL (14.12 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES O=C(OC)NC1=NC2=CC=C(C(C3=CC=CC=C3)=O)C=C2N1
Molecular Weight (g/mol) 295.29
Formula Weight 295.29
Percent Purity 99.34%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.