Learn More
BINDER™ Measurement of Air Exchange Rate
Product Code. 17827118 Shop All BINDER Products
Click to view available options
Service Offering:
Performance Test
Unit Size:
Each
Product Code. 17827118
Brand: BINDER™ 80122329
Only null left
Add to basket
Add to basket
Measurement of air exchange rate - in accordance with ASTM D5374, definition and protocol according to ambient temperature.
  • Accessory
Specifications
For Use With (Equipment) BINDER Units M 56, M 115, M 260, M 720
Certifications/Compliance ASTM D5374
Duration 1 Day
Product Type Test Report
Service Offering Performance Test
For Use With (Application) Measurement of Air Exchange Rate