Learn More
Invitrogen™ Measles virus Monoclonal Antibody (6017)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547565
Description
Specificity: Specific for Measles virus. These Mabys do not cross react with: Mumps virus, RSV, Para 1, Para 2 & Para 3. Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
Measles morbillivirus, formerly measles virus (MeV), is a single-stranded, negative-sense, enveloped, non-segmented RNA virus of the genus Morbillivirus within the family Paramyxoviridae. It is the cause of measles. Humans are the natural hosts of the virus; no animal reservoirs are known to exist.
Specifications
| Measles virus | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Proprietary sequence. | |
| 100 μg | |
| Primary | |
| 4°C or -20°C if preferred | |
| Liquid |
| ELISA, Immunocytochemistry | |
| 6017 | |
| Unconjugated | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.