Learn More
ME3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558408
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DVSLRIAIKV LDYAYKHNLA SYYPEPKDKE AFVRSLVYTP DYDSFTLDSY TWPKEAMNVQ TV Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 76%.
ME3 proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis, metastasis, and atherosclerosis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases.Specifications
ME3 | |
Polyclonal | |
Unconjugated | |
ME3 | |
1700020C08Rik; B230207H15Rik; HME; Macrophage elastase; malate dehydrogenase; malate dehydrogenase (oxaloacetate-decarboxylating) (NADP(+)); malic enzyme 3; malic enzyme 3, NADP(+)-dependent, mitochondrial; malic enzyme, NADP+-dependent, mitochondrial; ME; Me3; mitochondrial malic enzyme 3; mitochondrial NADP(+)-dependent malic enzyme 3; MME; MMP3; NADP-dependent malic enzyme, mitochondrial; NADP-ME; pyruvic-malic carboxylase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10873 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q16798 | |
ME3 | |
Recombinant protein corresponding to Human ME3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only