missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ME3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539278
This item is not returnable.
View return policy
Description
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis, metastasis, and atherosclerosis. Most MMP′s are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases.
Specifications
| ME3 | |
| Polyclonal | |
| Unconjugated | |
| ME3 | |
| Macrophage Metaloelastase, Macrophage Metaloelastase, Matrix Metalloprotease 3, Matrix Metalloprotease 3, MME, MME, MMP3, , HME, Macrophage elastase, ME | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10873, 109264 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q16798, Q8BMF3 | |
| ME3 | |
| A synthetic peptide derived from the internal region of human ME3 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction