missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: DGRVFTPGQG NNVYIFPGVA LAVILCNTRH ISDSVFLEAA KALTSQLTDE ELAQGRLYPP LANIQEVSIN IAIKVTEYLY ANKMAFRYPE PEDKAKYVKE RTWRSEYDSL LPDVYEWPES ASSPPV Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 87%.
ME2 encodes a mitochondrial NAD-dependent malic enzyme, a homotetrameric protein, that catalyzes the oxidative decarboxylation of malate to pyruvate. It had previously been weakly linked to a syndrome known as Friedreich ataxia that has since been shown to be the result of mutation in a completely different gene. Certain single-nucleotide polymorphism haplotypes of this gene have been shown to increase the risk for idiopathic generalized epilepsy. Alternatively spliced transcript variants encoding different isoforms found for this gene.
Specifications
Specifications
| Antigen | ME2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Me2 |
| Gene Accession No. | P23368 |
| Gene Alias | AW120568; D030040L20Rik; malate dehydrogenase; malate dehydrogenase (oxaloacetate-decarboxylating); malic enzyme 2; malic enzyme 2, NAD(+)-dependent, mitochondrial; Me2; NAD-dependent malic enzyme, mitochondrial; NAD-ME; ODS1; pyruvic-malic carboxylase |
| Gene Symbols | Me2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
