Learn More
ME2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24944-1-AP-20UL
 View more versions of this product
Product Code. 16883465
  • £135.00 / 20µL
Description
ME2 encodes a mitochondrial NAD-dependent malic enzyme, a homotetrameric protein, that catalyzes the oxidative decarboxylation of malate to pyruvate. It had previously been weakly linked to a syndrome known as Friedreich ataxia that has since been shown to be the result of mutation in a completely different gene. Certain single-nucleotide polymorphism haplotypes of this gene have been shown to increase the risk for idiopathic generalized epilepsy. Alternatively spliced transcript variants encoding different isoforms found for this gene.
Specifications
Specifications
ME2
Polyclonal
Unconjugated
ME2
Malic enzyme 2, ME2, NAD ME
Rabbit
Antigen Affinity Chromatography
RUO
4200
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P23368
ME2
ME2 Fusion Protein Ag20741
20 μL