missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ME2 Monoclonal Antibody (970CT2.1.1.2)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA537745
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
ME2 Monoclonal Antibody for Western Blot, IHC (P)
ME2 encodes a mitochondrial NAD-dependent malic enzyme, a homotetrameric protein, that catalyzes the oxidative decarboxylation of malate to pyruvate. It had previously been weakly linked to a syndrome known as Friedreich ataxia that has since been shown to be the result of mutation in a completely different gene. Certain single-nucleotide polymorphism haplotypes of this gene have been shown to increase the risk for idiopathic generalized epilepsy. Alternatively spliced transcript variants encoding different isoforms found for this gene.
Spezifikation
| ME2 | |
| Monoclonal | |
| 0.4 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P23368, Q99KE1 | |
| Me2 | |
| KLH conjugated synthetic peptide between 522-556 amino acids from the C-terminal region of human ME2. | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| 970CT2.1.1.2 | |
| Unconjugated | |
| Me2 | |
| AW120568; D030040L20Rik; malate dehydrogenase; malate dehydrogenase (oxaloacetate-decarboxylating); malic enzyme 2; malic enzyme 2, NAD(+)-dependent, mitochondrial; Me2; NAD-dependent malic enzyme, mitochondrial; NAD-ME; ODS1; pyruvic-malic carboxylase | |
| Mouse | |
| Protein G | |
| RUO | |
| 107029, 4200 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur