missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ME1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16619-1-AP-150UL
This item is not returnable.
View return policy
Description
ME1 encodes a cytosolic, NADP-dependent enzyme that generates NADPH for fatty acid biosynthesis. The activity of this enzyme, the reversible oxidative decarboxylation of malate, links the glycolytic and citric acid cycles. The regulation of expression for this gene is complex. Increased expression can result from elevated levels of thyroid hormones or by higher proportions of carbohydrates in the diet.Specifications
| ME1 | |
| Polyclonal | |
| Unconjugated | |
| ME1 | |
| HUMNDME, Malic enzyme 1, ME1, MES, NADP dependent malic enzyme, NADP ME | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17436, 24552, 4199 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P06801, P13697, P48163 | |
| Me1 | |
| ME1 Fusion Protein Ag9916 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction