missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ME1 Monoclonal Antibody (3I6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA523524
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, and HeLa cells. Predicted reactivity: Mouse (91%), Rat (91%), Chicken (80%), Sheep (92%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ME1 encodes a cytosolic, NADP-dependent enzyme that generates NADPH for fatty acid biosynthesis. The activity of this enzyme, the reversible oxidative decarboxylation of malate, links the glycolytic and citric acid cycles. The regulation of expression for this gene is complex. Increased expression can result from elevated levels of thyroid hormones or by higher proportions of carbohydrates in the diet.
Specifications
| ME1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P06801, P48163 | |
| Me1 | |
| Recombinant protein encompassing a sequence within the center region of human ME1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| 3I6 | |
| Unconjugated | |
| Me1 | |
| cytosolic malic enzyme 1; D9Ertd267e; HUMNDME; malate dehydrogenase; malate dehydrogenase (oxaloacetate-decarboxylating) (NADP(+)); malate dehydrogenase (oxaloacetate-decarboxylating) (NADP+); malate dehydrogenase decarboxylase (NADP+); malic enzyme; malic enzyme 1; malic enzyme 1, NADP(+)-dependent, cytoplasmic (cytosolic, soluble); malic enzyme 1, NADP(+)-dependent, cytosolic; malic enzyme 1, soluble; Malic enzyme, cytoplasmic; malic enzyme, supernatant; Mdh-1; ME1; MES; Mod1; Mod-1; NADP-dependent malic enzyme; NADP-dependent malic enzyme-like protein; NADP-ME; pyruvic-malic carboxylase | |
| Mouse | |
| Protein A | |
| RUO | |
| 17436, 4199 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction