missing translation for 'onlineSavingsMsg'
Learn More

ME1 Antibody (2A2), Novus Biologicals™

Product Code. 18389837 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389837 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18389837 Supplier Novus Biologicals Supplier No. H00004199M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ME1 Monoclonal antibody specifically detects ME1 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ME1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2A2
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH25246
Gene Alias EC 1.1.1, EC 1.1.1.40, HUMNDME, malate dehydrogenase, Malic enzyme 1, malic enzyme 1, NADP(+)-dependent, cytosolic, malic enzyme 1, soluble, Malic enzyme, cytoplasmic, MES, NADP-dependent malic enzyme, NADP-ME, pyruvic-malic carboxylase
Host Species Mouse
Immunogen ME1 (AAH25246, 1 a.a. ~ 572 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MEPEAPRRRHTHQRGYLLTRNPHLNKDLAFTLEERQQLNIHGLLPPSFNSQEIQVLRVVKNFEHLNSDFDRYLLLMDLQDRNEKLFYRVLTSDIEKFMPIVYTPTVGLACQQYSLVFRKPRGLFITIHDRGHIASVLNAWPEDVIKAIVVTDGERILGLGDLGCNGMGIPVGKLALYTACGGMNPQECLPVILDVGTENEELLKDPLYIGLRQRRVRGSEYDDFLDEFMEAVSSKYGMNCLIQFEDFANVNAFRLLNKYRNQYCTFNDDIQGTASVAVAGLLAALRITKNKLSDQTILFQGAGEAALGIAHLIVMALEKEGLPKEKAIKKIWLVDSKGLIVKGRASLTQEKEKFAHEHEEMKNLEAIVQEIKPTALIGVAAIGGAFSEQILKDMAAFNERPIIFALSNPTSKAECSAEQCYKITKGRAIFASGSPFDPVTLPNGQTLYPGQGNNSYVFPGVALGVVACGLRQITDNIFLTTAEVIAQQVSDKHLEEGRLYPPLNTIRDVSLKIAEKIVKDAYQEKTATVYPEPQNKEAFVRSQMYSTDYDQILPDCYSWPEEVQKIQTKVDQ
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4199
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.