missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDS028 Antibody (OTI3C4), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72614PCP
This item is not returnable.
View return policy
Description
MDS028 Monoclonal antibody specifically detects MDS028 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| MDS028 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 55846 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
| OTI3C4 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| integrin alpha FG-GAP repeat containing 2, integrin-alpha FG-GAP repeat-containing protein 2, MDS028 | |
| Full length human recombinant protein of human ITFG2 (NP_060933) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction