missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDS028 Antibody (OTI3C4), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72614MFV500
This item is not returnable.
View return policy
Description
MDS028 Monoclonal antibody specifically detects MDS028 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| MDS028 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 55846 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| OTI3C4 | |
| Flow Cytometry | |
| integrin alpha FG-GAP repeat containing 2, integrin-alpha FG-GAP repeat-containing protein 2, MDS028 | |
| Full length human recombinant protein of human ITFG2 (NP_060933) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction