missing translation for 'onlineSavingsMsg'
Learn More

MDS028 Antibody (1A8), Novus Biologicals™

Product Code. 18331619 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18331619 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18331619 Supplier Novus Biologicals Supplier No. H00055846M01100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MDS028 Monoclonal antibody specifically detects MDS028 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MDS028
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1A8
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH06552
Gene Alias integrin alpha FG-GAP repeat containing 2, integrin-alpha FG-GAP repeat-containing protein 2, MDS028
Host Species Mouse
Immunogen MDS028 (AAH06552, 1 a.a. ~ 447 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MRSVSYVQRVALEFSGSLFPHAICLGDVDNDTLNELVVGDTSGKVSVYKNDDSRPWLTCSCQGMLTCVGVGDVCNKGKNLLVAVSAEGWFHLFDLTPAKVLDASGHHETLIGEEQRPVFKQHIPANTKVMLISDIDGDGCRELVVGYTDRVVRAFRWEELGEGPEHLTGQLVSLKKWMLEGQVDSLSVTLGPLGLPELMVSQPGCAYAILLCTWKKDTGSPPASEGPTDGSRETPAARDVVLHQTSGRIHNKNVSTHLIGNIKQGHGTESSGSGLFALCTLDGTLKLMEEMEEADKLLWSVQVDHQLFALEKLDVTGNGHEEVVACAWDGQTYIIDHNRTVVRFQVDENIRAFCAGLYACKEGRNSPCLVYVTFNQKIYVYWEVQLERMESTNLVKLLETKPEYHSLLQELGVDPDDLPVTRALLHQTLYHPDQPPQCAPSSLQDPT
Purification Method IgG purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55846
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.