missing translation for 'onlineSavingsMsg'
Learn More

MDR-1339, MedChemExpress

Product Code. 16496078
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496078 10 mg 10mg
16482217 50 mg 50mg
16466476 100 mg 100mg
16463426 25 mg 25mg
16438687 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16496078 Supplier Medchem Express Supplier No. HY14503
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress MDR-1339 (DWK-1339) is an orally active and blood-brain-barrier-permeable Aβ-aggregation inhibitor, used in the research of Alzheimer's disease.

MDR-1339 (DWK-1339) is an orally active and blood-brain-barrier-permeable Aβ-aggregation inhibitor, used in the research of Alzheimer's disease.

Specifications

Chemical Name or Material MDR-1339
CAS 1018946-38-7
Color White
Purity Grade Notes Research
Molecular Formula C20H22O4
Quantity 10 mg
Synonym DWK-1339
Solubility Information DMSO : 50 mg/mL (153.19 mM; Need ultrasonic)
SMILES COCCCC1=CC=C(OC(C2=CC=C(OC)C(OC)=C2)=C3)C3=C1
Molecular Weight (g/mol) 326.39
Formula Weight 326.39
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.