missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MDP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551812
This item is not returnable.
View return policy
Description
Immunogen sequence: PKLAVFDLDY TLWPFWVDTH VDPPFHKSSD GTVRDRRGQD VRLYPEVPEV LKRLQSLGVP GAAASRTSEI EGANQLLELF DLFRYFVHRE IYPGSKITHF ERLQQKTGIP FSQMIFFDDE RRNIVDVSKL GVTCIHIQNG MNLQT Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 88%.
Magnesium-dependent phosphatase which may act as a tyrosine phosphatase.
Specifications
| MDP1 | |
| Polyclonal | |
| Unconjugated | |
| MDP1 | |
| 1810034K20Rik; AI035604; FN6PASE; fructosamine-6-phosphatase; magnesium dependent phosphatase 1; magnesium-dependent phosphatase 1; MDP1; MDP-1; RGD1311147 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 145553 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86V88 | |
| MDP1 | |
| Recombinant protein corresponding to Human MDP1. Recombinant protein control fragment (Product #RP-89041). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction