missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDM2(phospho S166) & MDM2 Protein Phosphorylation Antibody Pair, Abnova™
Antibody Pair
Brand: Abnova DP0097.1Set
This item is not returnable.
View return policy
Description
This protein phosphorylation antibody pair set comes with two antibodies, one against the MDM2 protein, and the other against the specific S166 phosphorylated site of MDM2 for use in in situ Proximity Ligation Assay.Specifications
| MDM2 | |
| Unconjugated | |
| HDMX/MGC71221/hdm2 | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Phospho-MDM2 S166 rabbit polyclonal antibody (20 ul) In PBS (0.09% (w/v) sodium azide) 2. MDM2 mouse monoclonal antibody (40 ug) In 1x PBS, pH 7.2 *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| In situ PLA | |
| Mdm2 p53 binding protein homolog (mouse) | |
| MDM2 | |
| RUO | |
| 4193 | |
| Human | |
| Ab Pair |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction