missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MDM2/HDM2 Monoclonal antibody specifically detects MDM2/HDM2 in Human samples. It is validated for ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | MDM2/HDM2 |
| Applications | ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | MDM2/7061R |
| Conjugate | Alexa Fluor 700 |
| Dilution | ELISA, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ACTFS, HDM2, HDMX, MDM2 oncogene, E3 ubiquitin protein ligase |
| Host Species | Rabbit |
| Immunogen | Recombinant human MDM2/HDM2 protein fragment (around aa 126-254) (exact sequence is proprietary) (Uniprot: Q00987) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?