missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDM2/HDM2 Antibody (HDM2-323) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-1312
This item is not returnable.
View return policy
Description
MDM2/HDM2 Monoclonal specifically detects MDM2/HDM2 in Human, Mouse samples. It is validated for Western Blot, ELISA.
Specifications
| MDM2/HDM2 | |
| Monoclonal | |
| 2.0 mg/mL | |
| Western Blot 1-2 ug/ml, ELISA 1:100-1:2000 | |
| MDM2 | |
| Synthetic peptide conjugated to KLH, corresponding to amino acids 389-402 of Human MDM2. | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Hypoxia, Neuroscience, p53 Pathway, Tumor Suppressors | |
| 4193 | |
| Human, Mouse | |
| IgG2a |
| Western Blot, ELISA | |
| HDM2-323 | |
| Unconjugated | |
| ACTFS, HDM2, HDMX, MDM2 oncogene, E3 ubiquitin protein ligase | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| MDM2 (HDM2-323) | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction