missing translation for 'onlineSavingsMsg'
Learn More

MDM2/HDM2 Antibody (197702) [DyLight 350], Novus Biologicals™

Product Code. 30732201 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732201 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732201 Supplier Novus Biologicals Supplier No. FAB1244D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MDM2/HDM2 Monoclonal antibody specifically detects MDM2/HDM2 in Human samples. It is validated for Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen MDM2/HDM2
Applications Immunoprecipitation
Classification Monoclonal
Clone 197702
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias ACTFS, HDM2, HDMX, MDM2 oncogene, E3 ubiquitin protein ligase
Host Species Mouse
Immunogen E. coli-derived recombinant human MDM2/HDM2, Asn3-Pro491, Accession # Q00987
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Cellular Markers, Hypoxia, Neuroscience, p53 Pathway, Phospho Specific, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4193
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.