missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16171-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a nuclear protein similar to the mouse double minute 1 protein. The mouse gene is located in double minute (DM) chromatin particles, is amplified in the mouse transformed 3T3 cell line, and the encoded protein is able to bind to p53. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| MDM1 | |
| Polyclonal | |
| Unconjugated | |
| MDM1 | |
| MDM1, Nuclear protein MDM1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17245, 314859, 56890 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5PQN4, Q8TC05, Q9D067 | |
| Mdm1 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction