Learn More
Tocris Bioscience™ MDL 72527
Polyamine oxidase (PAO) inhibitor
Description
Description
Polyamine oxidase (POA) inhibitor. Does not inhibit monoamine oxidase or D-Amino acid oxidase. Displays anticancer and neuroprotective activity in vivo.