missing translation for 'onlineSavingsMsg'
Learn More

MDL-29951, MedChemExpress

Product Code. 16463077
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463077 50 mg 50mg
16457086 100 mg 100mg
16447028 10 mg 10mg
16409577 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16463077 Supplier Medchem Express Supplier No. HY16312

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MDL-29951 is a novel glycine antagonist of NMDA receptor activation, with Ki of 0.14 μM for [3H]glycine binding in vitro and in vivo.

MDL-29951 is a novel glycine antagonist of NMDA receptor activation, with Ki of 0.14 μM for [3H]glycine binding in vitro and in vivo.

Specifications

Chemical Name or Material MDL-29951
CAS 130798-51-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C12H9Cl2NO4
Quantity 50 mg
Solubility Information DMSO : ≥ 50 mg/mL (165.50 mM)
SMILES O=C(O)CCC1=C(C(O)=O)NC2=C1C(Cl)=CC(Cl)=C2
Molecular Weight (g/mol) 302.11
Formula Weight 302.11
Percent Purity 99.5%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.