missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ MDL 11,939
GREENER_CHOICE

Product Code. 30001436
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001436 10 mg 10mg
18045392 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001436 Supplier Tocris Bioscience™ Supplier No. 0870/10
  • ONLINE EXCLUSIVE PRICE
    £183.00
    £172.20 / 10mg
    Save £10.80
    5% Off
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

5-HT2A antagonist

Orally active 5-HT2A receptor antagonist; displays selectivity for 5-HT2A receptors over 5-HT2C receptors (Ki values are 0.54, 2.5, 81.6 and ∽10,000 nM at rabbit 5-HT2A, human 5-HT2A, rabbit 5-HT2C and human 5-HT2C receptors respectively).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107703-78-6
Molecular Formula C20H25NO

Specifications

Quantity 10 mg
Formula Weight 295.42
Purity >99%
Molecular Weight (g/mol) 295.426
Chemical Name or Material MDL 11, 939
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.