missing translation for 'onlineSavingsMsg'
Learn More

MDL 105519, MedChemExpress

Product Code. 16496308
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496308 10 mg 10mg
16472427 50 mg 50mg
16448897 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16496308 Supplier Medchem Express Supplier No. HY15085

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MDL 105519 is a potent and selective antagonist of glycine binding to the NMDA receptor.

MDL 105519 is a potent and selective antagonist of glycine binding to the NMDA receptor.

Specifications

Chemical Name or Material MDL 105519
CAS 161230-88-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H11Cl2NO4
Quantity 10 mg
Solubility Information DMSO : 17 mg/mL (45.19 mM; Need ultrasonic and warming)
SMILES O=C(O)C(N1)=C(/C=C(C2=CC=CC=C2)/C(O)=O)C3=C1C=C(Cl)C=C3Cl
Molecular Weight (g/mol) 376.19
Formula Weight 376.19
Percent Purity 97.15%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.