missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDL-1/CLEC5A Antibody (283834) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2384JF585
This item is not returnable.
View return policy
Description
MDL-1/CLEC5A Monoclonal antibody specifically detects MDL-1/CLEC5A in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| MDL-1/CLEC5A | |
| Monoclonal | |
| Janelia Fluor 585 | |
| CLECSF5carbohydrate-recognition domain) lectin, superfamilymember 5, C-type lectin domain family 5, member A, C-type lectin superfamily member 5, MDL-1MGC138304, Myeloid DAP12-associating lectin 1, myeloid DAP12-associating lectin-1 | |
| Mouse myeloma cell line NS0-derived recombinant human MDL-1/CLEC5A, Tyr26-Lys188, Accession # Q9NY25 | |
| 0.1 mL | |
| Immunology | |
| 23601 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Western Blot, CyTOF | |
| 283834 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction