missing translation for 'onlineSavingsMsg'
Learn More

MDH2 Antibody (2B7), Novus Biologicals™

Product Code. 18394568 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394568 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18394568 Supplier Novus Biologicals Supplier No. H00004191M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MDH2 Monoclonal antibody specifically detects MDH2 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MDH2
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2B7
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_005909
Gene Alias EC 1.1.1, EC 1.1.1.37, malate dehydrogenase 2, NAD (mitochondrial), MDH, MGC:3559, mitochondrial, MOR1
Host Species Mouse
Immunogen MDH2 (NP_005909, 134 a.a. ~ 246 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EAMICVIANPVNSTIPITAEVFKKHGVYNPNKIFGVTTLDIVRANTFVAELKGLDPARVNVPVIGGHAGKTIIPLISQCTPKVDFPQDQLTALTGRIQEAGTEVVKAKAGAGS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4191
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.