missing translation for 'onlineSavingsMsg'
Få mere at vide

MDH1B Antibody, Novus Biologicals™

Artikelnummer. 18415619 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
0.05 mg
Pakningsstørrelse:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
18415619 0.05 mg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18415619 Leverandør Novus Biologicals Leverandørnr. H00130752B01P

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Mouse Polyclonal Antibody

MDH1B Polyclonal antibody specifically detects MDH1B in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen MDH1B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. NP_001034934.1
Gene Alias EC 1.1.1.-, FLJ25341, malate dehydrogenase 1B, NAD (soluble), putative malate dehydrogenase 1B, RP11-95H11
Host Species Mouse
Immunogen MDH1B (NP_001034934.1, 1 a.a. - 518 a.a.) full-length human protein. MAKFVIAGRADCPYYAKTELVADYLQKNLPDFRIHKITQRPEVWEDWLKDVCEKNKWSHKNSPIIWRELLDRGGKGLLLGGYNEFLEHAQLYYDVTSSMTTELMMVIAQENLGAHIEKEQEEEALKTCINPLQVWITSASAPACYNLIPILTSGEVFGMHTEISITLFDNKQAEEHLKSLVVETQDLASPVLRSVSICTKVEEAFRQAHVIVVLDDSTNKEVFTLEDCLRSRVPLCRLYGYLIEKNAHESVRVIVGGRTFVNLKTVLLMRYAPRIAHNIIAVALGVEGEAKAILARKLKTAPSYIKDVIIWGNISGNNYVDLRKTRVYRYESAIWGPLHYSRPVLNLIFDSEWVKREFVAILKNLTTTGRQFGGILAAHSIATTLKYWYHGSPPGEIVSLGILSEGQFGIPKGIVFSMPVKFENGTWVVLTDLKDVEISEQIMTRMTSDLIQEKLVALGDKIHFQPYQSGHKDLVPDEEKNLAMSDAAEFPNQIPQTTFEKPQSLEFLNEFEGKTVES
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 130752
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.