missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDH1B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | MDH1B |
|---|---|
| Dilution | Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18643478
|
Novus Biologicals
NBP2-68969-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18608016
|
Novus Biologicals
NBP2-68969 |
100 μg |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MDH1B Polyclonal antibody specifically detects MDH1B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| MDH1B | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.2) and 40% Glycerol | |
| 130752 | |
| IgG | |
| Protein A purified |
| Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 1.1.1.-, FLJ25341, malate dehydrogenase 1B, NAD (soluble), putative malate dehydrogenase 1B, RP11-95H11 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MAKFVIAGRADCPYYAKTELVADYLQKNLPDFRIHKITQRPEVWEDWLKDVCEKNKWSHKNSPIIWRELLDR | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title