missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MDH1 Recombinant Rabbit Monoclonal Antibody (JE35-03)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556543
This item is not returnable.
View return policy
Description
MDH1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
MDH1 is localized to the cytoplasm and may play pivotal roles in the malate-aspartate shuttle that operates in the metabolic coordination between cytosol and mitochondria.
Specifications
| MDH1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O88989, P14152, P40925 | |
| MDH1 | |
| Synthetic peptide within human mdh1 aa 285-334/334. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| JE35-03 | |
| Unconjugated | |
| MDH1 | |
| B230377B03Rik; Cytosolic malate dehydrogenase; D17921; Diiodophenylpyruvate reductase; epididymis secretory protein Li 32; HEL-S-32; malate dehydrogenase (EC 1.1.1.37); malate dehydrogenase 1; malate dehydrogenase 1, NAD (soluble); malate dehydrogenase, cytoplasmic; malate dehydrogenase, cytoplasmic; malate dehydrogenase, peroxisomal; malate dehydrogenase, soluble; Malate dehydrogenase-like enzyme; Mdh; Mdh1; MDHA; Mdhl; MDH-s; MDL1; MGC:1375; Mor2; Mor-2; soluble malate dehydrogenase | |
| Rabbit | |
| Protein A | |
| RUO | |
| 17449, 24551, 4190 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction