missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDH1 Rabbit anti-Human, Mouse, Rat, Clone: 8U9X1, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | MDH1 |
|---|---|
| Clone | 8U9X1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18319765
|
Novus Biologicals
NBP3-16860-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18396744
|
Novus Biologicals
NBP3-16860-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MDH1 Monoclonal antibody specifically detects MDH1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| MDH1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Immunology, Stem Cell Markers, Virology Bacteria and Parasites | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 4190 | |
| IgG | |
| Affinity purified |
| 8U9X1 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| cytoplasmic, Cytosolic malate dehydrogenase, DEE88, Diiodophenylpyruvate reductase, EC 1.1.1.37, EC 1.1.1.96, HEL-S-32, malate dehydrogenase 1, NAD (soluble), Malate dehydrogenase, cytoplasmic, MDH1, MDHA, MDH-s, MGC:1375, MOR2, soluble malate dehydrogenase | |
| A synthetic peptide corresponding to a sequence within amino acids 235-334 of human MDH1 (P40925). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title