missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDH1 Mouse anti-Human, Mouse, Clone: 1F9A2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66505-1-IG-20UL
This item is not returnable.
View return policy
Description
MDH1 is localized to the cytoplasm and may play pivotal roles in the malate-aspartate shuttle that operates in the metabolic coordination between cytosol and mitochondria.Specifications
| MDH1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P14152, P40925 | |
| Mdh1 | |
| MDH1 Fusion Protein Ag8744 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 1F9A2 | |
| Unconjugated | |
| MDH1 | |
| Cytosolic malate dehydrogenase, MDH s, MDH1, MDHA, MOR2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 17449, 4190 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction