missing translation for 'onlineSavingsMsg'
Learn More

MDH1 Antibody, PE, Novus Biologicals™

Product Code. 18416880 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416880 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416880 Supplier Novus Biologicals Supplier No. NBP177780PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

MDH1 Polyclonal antibody specifically detects MDH1 in Porcine samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen MDH1
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias cytoplasmic, Cytosolic malate dehydrogenase, DEE88, Diiodophenylpyruvate reductase, EC 1.1.1.37, EC 1.1.1.96, HEL-S-32, malate dehydrogenase 1, NAD (soluble), Malate dehydrogenase, cytoplasmic, MDH1, MDHA, MDH-s, MGC:1375, MOR2, soluble malate dehydrogenase
Host Species Sheep
Immunogen MDH1 [Pig Heart] (Uniprot: P11708)
Purification Method Multi-step
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology, Stem Cell Markers, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 4190
Target Species Pig
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.