missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDH1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NBP1-77780AF750
This item is not returnable.
View return policy
Description
MDH1 Polyclonal antibody specifically detects MDH1 in Porcine samples. It is validated for Western Blot, ELISA
Specifications
| MDH1 | |
| Polyclonal | |
| Western Blot, ELISA | |
| cytoplasmic, Cytosolic malate dehydrogenase, DEE88, Diiodophenylpyruvate reductase, EC 1.1.1.37, EC 1.1.1.96, HEL-S-32, malate dehydrogenase 1, NAD (soluble), Malate dehydrogenase, cytoplasmic, MDH1, MDHA, MDH-s, MGC:1375, MOR2, soluble malate dehydrogenase | |
| MDH1 [Pig Heart] (Uniprot: P11708) | |
| 0.1 mL | |
| Cancer, Immunology, Stem Cell Markers, Virology Bacteria and Parasites | |
| 4190 | |
| Store at 4C in the dark. |
| Western Blot, ELISA | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Sheep | |
| Multi-step | |
| RUO | |
| Primary | |
| Pig | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction