missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: ALVQLIVQYP PAVEPAFLEI RQGQDRSVTM SCRVLRAYPI RVLTYEWRLG NKLLRTGQFD SQEYTEYAVK SLSNENYGVY NCSIINEAGA GRCSFLVTGK AYAPEFYYDT YNPVWQNRHR VYSYSLQWTQ MNPDAV Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
MDGA2 may involved in cell-cell interactions.
Specifications
Specifications
| Antigen | MDGA2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MDGA2 |
| Gene Accession No. | Q7Z553 |
| Gene Alias | 6720489L24Rik; 9330209L04Rik; acrodysplasia; Adp; c14_5286; MAM domain containing 1; MAM domain containing glycosylphosphatidylinositol anchor 2; MAM domain-containing glycosylphosphatidylinositol anchor protein 2; MAM domain-containing protein 1; MAMDC1; Mdga2; UNQ8188/PRO23197 |
| Gene Symbols | MDGA2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
