Learn More
MDGA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556686
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: AHVPISPSGP FQIIFEGVRG PGYLGDIAID DVTLKKGECP RKQTDPNKVV VMPGSGAPCQ SSPQLWGPMA IFLLALQR Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 88%.
MDGA1 Polyclonal antibody specifically detects MDGA1 in Human samples. It is validated for Immunohistochemistry.Specifications
MDGA1 | |
Polyclonal | |
Unconjugated | |
MDGA1 | |
FLJ45018; glycosyl-phosphatidyl-inositol-MAM; glycosylphosphatidylinositol-MAM; GPI and MAM protein; GPIM; MAM domain containing glycosylphosphatidylinositol anchor 1; MAM domain-containing glycosylphosphatidylinositol anchor protein 1; MAM domain-containing protein 3; MAMDC3; MDGA1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
266727 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NFP4 | |
MDGA1 | |
Recombinant protein corresponding to Human MDGA1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only