missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDFI Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35380-20ul
This item is not returnable.
View return policy
Description
MDFI Polyclonal antibody specifically detects MDFI in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| MDFI | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| I-MF, I-mfa, inhibitor of MyoD family a, MyoD family inhibitor, Myogenic repressor I-mf | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human MDFI (NP_005577.1).,, Sequence:, MYQVSGQRPSGCDAPYGAPSAAPGPAQTLSLLPGLEVVTGSTHPAEAAPEEGSLEEAATPMPQGNGPGIPQGLDSTDLDVPTEAVTCQPQGNPLGCTPLLPNDSGHPSELGGTRRAGNGALGGPKAHRKLQTHPSLASQGSKKSKSSSKSTTSQIPLQAQ | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 4188 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction