missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDFI Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-49886
This item is not returnable.
View return policy
Description
MDFI Polyclonal specifically detects MDFI in Human samples. It is validated for Peptide ELISA.
Specifications
| MDFI | |
| Polyclonal | |
| Peptide ELISA Detection limit 1:4000 | |
| NP_005577.1 | |
| MDFI | |
| Peptide with sequence YQVSGQRPSGCD corresponding to N-Terminus according to NP_005577.1. | |
| 0.1 mg | |
| Primary | |
| This antibody is specific to the N Terminus. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| I-MF, I-mfa, inhibitor of MyoD family a, MyoD family inhibitor, Myogenic repressor I-mf | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4188 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction