missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDC1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-395CL1
This item is not returnable.
View return policy
Description
MDC1 Polyclonal antibody specifically detects MDC1 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| MDC1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| DKFZp781A0122, Em:AB023051.5, KIAA0170MGC166888, mediator of DNA damage checkpoint 1, mediator of DNA damage checkpoint protein 1, mediator of DNA-damage checkpoint 1, NFBD1homologue to Drosophila photoreceptor protein calphotin, Nuclear factor with BRCT domains 1 | |
| A synthetic peptide, which represented a portion of human Mediator of DNA Damage Checkpoint Protein 1 encoded within exon 5 (NP_055456.1). | |
| 0.1 mL | |
| Autophagy, DNA Repair | |
| 9656 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction