missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MDA5 Recombinant Rabbit Monoclonal Antibody (23GB6270), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553011
This item is not returnable.
View return policy
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp and a protein kinase C-activating compound, mezerein. Irreversible reprogramming of melanomas can be achieved by treatment with both these agents; treatment with either agent alone only achieves reversible differentiation.
Specifications
| MDA5 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| IFIH1 | |
| 9130009C22Rik; AGS7; CADM-140 autoantigen; Clinically amyopathic dermatomyositis autoantigen 140 kDa; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide; helicard; Helicase with 2 CARD domains; Hlcd; IDDM19; IFIH1; interferon induced with helicase C domain 1; interferon induced with helicase C domain protein 1; interferon induced, with helicase C domain 1; interferon-induced helicase C domain-containing protein 1; Interferon-induced with helicase C domain protein 1; MDA5; MDA-5; melanoma differentiation associated protein-5; melanoma differentiation-associated gene 5; melanoma differentiation-associated protein 5; MGC133047; Murabutide down-regulated protein; RGD1560496; RH116; RIG-I-like receptor 2; RLR-2; RNA helicase-DEAD box protein 116; SGMRT1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 499801, 64135, 71586 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 23GB6270 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8R5F7, Q9BYX4 | |
| IFIH1 | |
| A synthesized peptide derived from human MDA5 (950-1025AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction