missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDA5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £435.00
Specifications
| Antigen | MDA5 |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18440841
|
Novus Biologicals
NBP1-89685-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18465051
|
Novus Biologicals
NBP1-89685 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MDA5 Polyclonal antibody specifically detects MDA5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| MDA5 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS (pH 7.2) and 40% Glycerol | |
| 64135 | |
| IgG | |
| Immunogen affinity purified |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CADM-140 autoantigen, Clinically amyopathic dermatomyositis autoantigen 140 kDa, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide, EC 3.6.4.13, Helicard, Helicase with 2 CARD domains, Hlcd, IDDM19, interferon induced with helicase C domain 1, interferon-induced helicase C domain-containing protein 1, Interferon-induced with helicase C domain protein 1, MDA-5MGC133047, MDA5RNA helicase-DEAD box protein 116, melanoma differentiation associated protein-5, Melanoma differentiation-associated protein 5, Murabutide down-regulated protein, RH116 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: TIRMIDAYTHLETFYNEEKDKKFAVIEDDSDEGGDDEYCDGDEDEDDLKKPLKLDETDRFLMTLFFENNKMLKRLAENPEYENEKLTKLRNTIMEQYTRTEESARGIIFTKTRQSAYALSQWITENEKFAEVGVKAHHLIGAGHSSEFKP | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title