missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ MDA-MB-231 Whole Cell Lysate

Product Code. 18370772 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370772 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18370772 Supplier Novus Biologicals™ Supplier No. NBP318060

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

for use as a positive control in Western Blot

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) This product has been tested by SDS-PAGE. Multi-purpose MDA-MB-231 Whole Cell Lysate are especially prepared as positive control for multiple assays including western blot, immunoprecipitation (IP), capture ELISA or other assays requiring native protein sample. For separation by SDS-PAGE and subsequent western blot analysis, lysates should be diluted by user to desired concentration in SDS-PAGE buffer with 2-mercaptoethanol or dithiothreitol as the reducing agent and heated to 95C for 5 minutes. Sample is ready for use in immunoprecipitation and ELISA experiments, conditions should be optimized by the user. It is recommended to use TrueBlot IP reagents for immunoprecipitation experiments.
Quantity 500 μg
Content And Storage Store at -70C. Avoid freeze-thaw cycles.
Target MDA-MB-231
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.