missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDA 19, MedChemExpress
Description
MDA 19 is a potent and selective agonist of human cannabinoid receptor 2 (CB2), with a Ki of 43.3 nM. MDA 19 has antiallodynic effects in a rat model of neuropathic pain and does not affect rat locomotor activity.
Specifications
Specifications
| Chemical Name or Material | MDA 19 |
| Physical Form | Solid |
| CAS | 1048973-47-2 |
| Color | Yellow |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C21H23N3O2 |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Solubility Information | DMSO : 14.29 mg/mL (40.90 mM; Need ultrasonic) |
| SMILES | O=C(N/N=C1C(N(CCCCCC)C2=C\1C=CC=C2)=O)C3=CC=CC=C3 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?