missing translation for 'onlineSavingsMsg'
Learn More

MD-2 Antibody - BSA Free, Novus Biologicals™

Product Code. 18386710 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18386710 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18386710 Supplier Novus Biologicals Supplier No. NBP177201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MD-2 Polyclonal specifically detects MD-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MD-2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1 ug/mL, ELISA 1:100-1:2000, Immunohistochemistry 2-2.5 ug/mL, Immunocytochemistry/Immunofluorescence 10-20 ug/mL, Immunohistochemistry-Paraffin 2 ug/ml
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_056179
Gene Alias ESOP1, ESOP-1, Ly-96, lymphocyte antigen 96, MD2, MD-2, myeloid differentiation protein-2, Protein MD-2
Gene Symbols LY96
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 13 amino acids near the center of human MD-2. The immunogen is located within the last 50 amino acids of MD-2. Amino Acid Squence: CSFKGIKFSKGKYK
Purification Method Peptide affinity purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23643
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.