missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MD-1/LY86 Monoclonal specifically detects MD-1/LY86 in Mouse samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | MD-1/LY86 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 416 |
| Conjugate | DyLight 755 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Ly-86, lymphocyte antigen 86, MD1, MMD-1, Protein MD-1, RP105-associated |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Mouse MD-1/LY86 (Met 1-Ser 162; NP_034875.1). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?