missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCU Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120437
This item is not returnable.
View return policy
Description
Positive test controls include: HepG2. Immunogen sequence: VHQRIASWQN LGAVYCSTVV PSDDVTVVYQ NGLPVISVRL PSRRERCQFT LKPISDSVGV FLRQLQEEDR GIDRVAIYSP DGVRVAASTG IDLLLLDDFK LVINDLTYHV RPPKRDLLSH.
Mitochondrial inner membrane calcium uniporter that mediates calcium uptake into mitochondria. Constitutes the pore-forming and calcium-conducting subunit of the uniporter complex (uniplex). Activity is regulated by MICU1 and MICU2. At low Ca(2+) levels MCU activity is down-regulated by MICU1 and MICU2; at higher Ca(2+) levels MICU1 increases MCU activity. Mitochondrial calcium homeostasis plays key roles in cellular physiology and regulates cell bioenergetics, cytoplasmic calcium signals and activation of cell death pathways. Involved in buffering the amplitude of systolic calcium rises in cardiomyocytes. While dispensable for baseline homeostatic cardiac function, acts as a key regulator of short-term mitochondrial calcium loading underlying a 'fight-or-flight' response during acute stress: acts by mediating a rapid increase of mitochondrial calcium in pacemaker cells. participates in mitochondrial permeability transition during ischemia-reperfusion injury. Regulates glucose-dependent insulin secretion in pancreatic beta-cells by regulating mitochondrial calcium uptake. Mitochondrial calcium uptake in skeletal muscle cells is involved in muscle size in adults. Regulates synaptic vesicle endocytosis kinetics in central nerve terminal. Involved in antigen processing and presentation.
Specifications
| MCU | |
| Polyclonal | |
| Unconjugated | |
| MCU | |
| 2010012O16Rik; AV064928; C10orf42; Calcium uniporter protein, mitochondrial; Ccdc109a; coiled-coil domain containing 109A; coiled-coil domain-containing protein 109A; D130073L02Rik; Gm64; HsMCU; MCU; mitochondrial calcium uniporter | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 215999, 294560, 90550 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.07 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q3UMR5, Q8NE86 | |
| MCU | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction