missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCTS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14984-1-AP-150UL
This item is not returnable.
View return policy
Description
MCTS1 play a role in cell cycle regulation; decreases cell doubling time and anchorage-dependent growth; shortens the duration of G1 transit time and G1/S transition.Specifications
| MCTS1 | |
| Polyclonal | |
| Unconjugated | |
| MCTS1 | |
| MCT 1, MCT1, MCTS1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 28985, 302500, 68995 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q4G009, Q9DB27, Q9ULC4 | |
| MCTS1 | |
| MCTS1 Fusion Protein Ag6968 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction